Product Details

SNP ID
rs9829954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:13976783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAACCTAATCAAAGACTTTTCAC[A/G]AGTTAACAATTCTCATAGATTTCCA
Phenotype
MIM: 611167
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TPRXL PubMed Links

Gene Details

Gene
TPRXL
Gene Name
tetrapeptide repeat homeobox like
There are no transcripts associated with this gene.

View Full Product Details