Product Details

SNP ID
rs10439116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51220964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTATCTGATGAATGACTTGTATCC[A/T]GAATATATCAAGGACTCTCAAAACA
Phenotype
MIM: 159590
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CD33 PubMed Links

Gene Details

Gene
CD33
Gene Name
CD33 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082618.1 Intron NP_001076087.1
NM_001177608.1 Intron NP_001171079.1
NM_001772.3 Intron NP_001763.3
XM_011527531.2 Intron XP_011525833.1
XM_011527532.2 Intron XP_011525834.1
XM_017027508.1 Intron XP_016882997.1
XM_017027509.1 Intron XP_016882998.1
XM_017027510.1 Intron XP_016882999.1

View Full Product Details