Product Details

SNP ID
rs10174244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61150064 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATACTGGGTCTGCAAAGATCGCC[A/G]AATGGTTGATGGAAGAATACTATAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf74 PubMed Links

Gene Details

Gene
C2orf74
Gene Name
chromosome 2 open reading frame 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143959.1 Intron NP_001137431.1
NM_001143960.2 Intron NP_001137432.1
NM_001316317.1 Intron NP_001303246.1
XM_017003967.1 Intron XP_016859456.1
Gene
KIAA1841
Gene Name
KIAA1841
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005264605.3 Intron XP_005264662.1
XM_005264606.4 Intron XP_005264663.1
XM_005264607.2 Intron XP_005264664.1
XM_011533126.2 Intron XP_011531428.1
XM_011533127.2 Intron XP_011531429.1
XM_011533128.1 Intron XP_011531430.1
XM_011533129.2 Intron XP_011531431.1
XM_011533131.2 Intron XP_011531433.1
XM_017005108.1 Intron XP_016860597.1
XM_017005109.1 Intron XP_016860598.1
Gene
LOC339803
Gene Name
uncharacterized LOC339803
There are no transcripts associated with this gene.

View Full Product Details