Product Details

SNP ID
rs10199063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169482919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGTTGGAACTTTTAAAAGTCCA[A/G]GCTGCCCATAGCAGCTATCTGTGGA
Phenotype
MIM: 603650
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BBS5 PubMed Links

Gene Details

Gene
BBS5
Gene Name
Bardet-Biedl syndrome 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152384.2 Intron NP_689597.1

View Full Product Details