Product Details

SNP ID
rs9939297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67905391 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACTCTGGGTTTCTGAGCCTTTGC[A/G]GTGTGTTATTCTCTCTCCCTGAAAC
Phenotype
MIM: 177015
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSKH1 PubMed Links

Gene Details

Gene
PSKH1
Gene Name
protein serine kinase H1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006742.2 Intron NP_006733.1

View Full Product Details