Product Details

SNP ID
rs10405306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45093965 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAAAAAGAGGTTAAGTTGCTGGT[C/T]TGCAAGCCTTGGGGCAAGGAGTGGT
Phenotype
MIM: 607419
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GEMIN7 PubMed Links

Gene Details

Gene
GEMIN7
Gene Name
gem nuclear organelle associated protein 7
There are no transcripts associated with this gene.

Gene
LOC105372419
Gene Name
uncharacterized LOC105372419
There are no transcripts associated with this gene.

Gene
PPP1R37
Gene Name
protein phosphatase 1 regulatory subunit 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019121.1 Intron NP_061994.1
XM_006723159.2 Intron XP_006723222.1

View Full Product Details