Product Details

SNP ID
rs9771343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:20785535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGACACCAGGGCCGCGGCAGCCGC[A/G]GCTGCTGCCGCGGCCGCCGCAGCCG
Phenotype
MIM: 608306
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SP8 PubMed Links

Gene Details

Gene
SP8
Gene Name
Sp8 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182700.5 469 Silent Mutation GCC,GCT A94A NP_874359.2
NM_198956.3 469 Silent Mutation GCC,GCT A76A NP_945194.1

View Full Product Details