Product Details

SNP ID
rs9296115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34466938 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTACAAGAGGCTATGAAAAGGTTA[A/C]AGGGTCATCAGGGTGTAGGAGACAT
Phenotype
MIM: 606512
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PACSIN1 PubMed Links

Gene Details

Gene
PACSIN1
Gene Name
protein kinase C and casein kinase substrate in neurons 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199583.2 Intron NP_001186512.1
NM_020804.4 Intron NP_065855.1
XM_011514541.1 Intron XP_011512843.1

View Full Product Details