Product Details

SNP ID
rs9633990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66640691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTAGAATTACAAGGCTGATTTTA[C/G]GACTTCCTGGGGAGGAGGCACTGGG
Phenotype
MIM: 602571
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RBM14-RBM4 PubMed Links

Gene Details

Gene
RBM14-RBM4
Gene Name
RBM14-RBM4 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198845.1 Intron NP_001185774.1
NM_001198846.1 Intron NP_001185775.1
Gene
RBM4
Gene Name
RNA binding motif protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198843.1 Intron NP_001185772.1
NM_001198844.1 Intron NP_001185773.1
NM_002896.3 Intron NP_002887.2

View Full Product Details