Product Details
- SNP ID
-
rs6638329
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:135165893 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGAAGAAATGGAAAATCTGAACAG[A/C]CCATTAATGAGTAAGGAAAGTGATT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CT55
PubMed Links
Gene Details
- Gene
- CT55
- Gene Name
- cancer/testis antigen 55
View Full Product Details