Product Details

SNP ID
rs10190003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:186492738 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTACTATTGTTTTCTTTTTCTAG[C/T]TTGTCCACGTGGTGGTATTTTGTAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZC3H15 PubMed Links

Gene Details

Gene
ZC3H15
Gene Name
zinc finger CCCH-type containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018471.2 Intron NP_060941.2
XM_017004502.1 Intron XP_016859991.1

View Full Product Details