Product Details

SNP ID
rs7260025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16669813 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAGTGCCCTGGGGACCCTTCTAG[C/T]ACCACCCAGTCATCTGCAAATGAGG
Phenotype
MIM: 611235
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMIM7 PubMed Links

Gene Details

Gene
SMIM7
Gene Name
small integral membrane protein 7
There are no transcripts associated with this gene.

Gene
TMEM38A
Gene Name
transmembrane protein 38A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024074.2 Intron NP_076979.1

View Full Product Details