Product Details

SNP ID
rs7532933
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:64210769 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCCACACACTGGTCAGCCCTGTA[C/T]AGACTTTTTGGACAACCCTGAGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE2U PubMed Links

Gene Details

Gene
UBE2U
Gene Name
ubiquitin conjugating enzyme E2 U (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152489.1 1248 Missense Mutation ACA,ATA T90I NP_689702.1
XM_005270498.4 1248 Missense Mutation ACA,ATA T90I XP_005270555.1
XM_006710379.3 1248 Missense Mutation ACA,ATA T58I XP_006710442.1
XM_011540764.2 1248 Missense Mutation ACA,ATA T90I XP_011539066.1
XM_017000379.1 1248 Intron XP_016855868.1
XM_017000380.1 1248 Intron XP_016855869.1
XM_017000381.1 1248 Intron XP_016855870.1
XM_017000382.1 1248 Intron XP_016855871.1
XM_017000383.1 1248 Intron XP_016855872.1
XM_017000384.1 1248 Intron XP_016855873.1
XM_017000385.1 1248 Intron XP_016855874.1

View Full Product Details