Product Details

SNP ID
rs7643236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57268965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGAAGAATGCATTAAATGATCCA[A/T]GAAATTGTAAGAGCGTGTTATGGGT
Phenotype
MIM: 604299
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
APPL1 PubMed Links

Gene Details

Gene
APPL1
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012096.2 2561 Intron NP_036228.1
XM_011533583.2 2561 Intron XP_011531885.1
Gene
ASB14
Gene Name
ankyrin repeat and SOCS box containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142733.2 2561 UTR 3 NP_001136205.2
NM_130387.5 2561 UTR 3 NP_569058.1
XM_017005736.1 2561 Intron XP_016861225.1
XM_017005737.1 2561 Intron XP_016861226.1
Gene
LOC105377102
Gene Name
uncharacterized LOC105377102
There are no transcripts associated with this gene.

View Full Product Details