Product Details

SNP ID
rs6622268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:107838787 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTCTCCAATGATGACAAGATAAT[A/G]AACAAAAGCTACTCAAAAGCAAACA
Phenotype
MIM: 300204
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MID2 PubMed Links

Gene Details

Gene
MID2
Gene Name
midline 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012216.3 Intron NP_036348.2
NM_052817.2 Intron NP_438112.2
XM_005262062.4 Intron XP_005262119.1
XM_017029239.1 Intron XP_016884728.1

View Full Product Details