Product Details

SNP ID
rs10059907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:205245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGACGCCGGAGACCCAGAAGGC[C/G]CATGACTGCCTGGCCTCGAGTCACT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC127 PubMed Links

Gene Details

Gene
CCDC127
Gene Name
coiled-coil domain containing 127
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145265.2 935 UTR 3 NP_660308.1
Gene
LRRC14B
Gene Name
leucine rich repeat containing 14B
There are no transcripts associated with this gene.

View Full Product Details