Product Details

SNP ID
rs10443172
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:85020085 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCGAATAACACACAGTTCTCTGCA[C/T]GCAGAGTTGGAGGCAGTGAACTTTT
Phenotype
MIM: 607400
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCOLN3 PubMed Links

Gene Details

Gene
MCOLN3
Gene Name
mucolipin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253693.1 Intron NP_001240622.1
NM_018298.10 Intron NP_060768.8
XM_005271003.1 Intron XP_005271060.1
XM_006710750.1 Intron XP_006710813.1
XM_011541739.2 Intron XP_011540041.2
XM_011541740.2 Intron XP_011540042.1
XM_011541741.2 Intron XP_011540043.1
XM_011541742.1 Intron XP_011540044.1

View Full Product Details