Product Details

SNP ID
rs10207196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68043126 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACTGAATTTTATTATACTGTACA[C/T]CTTGGTTAAATTCAACATATATGTA
Phenotype
MIM: 606997
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1D PubMed Links

Gene Details

Gene
C1D
Gene Name
C1D nuclear receptor corepressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190263.1 Intron NP_001177192.1
NM_001190265.1 Intron NP_001177194.1
NM_006333.3 Intron NP_006324.1
NM_173177.2 Intron NP_775269.1

View Full Product Details