Product Details

SNP ID
rs10174299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:192759276 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAATTTTTACATTTTATAACAAAA[A/T]TCAAATTTAAAAATGAACAAGCAGT
Phenotype
MIM: 605443
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PCGEM1 PubMed Links

Gene Details

Gene
PCGEM1
Gene Name
PCGEM1, prostate-specific transcript (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details