Product Details
- SNP ID
-
rs9494888
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:99613722 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACCACCACGCGCACCACCACCAC[A/C]ATCCCAAGTGCCTGCTCGCTGGGGA
- Phenotype
-
MIM: 616741
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PRDM13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs577768379] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRDM13
- Gene Name
- PR domain 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021620.3 |
1348 |
Missense Mutation |
AAT,CAT |
N363H |
NP_067633.2 |
View Full Product Details