Product Details

SNP ID
rs4987858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63125561 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGCCAGTGTAAAGAGGAGTACAT[A/G]CAGAGGACTGTTTTTTCATTCATAA
Phenotype
MIM: 151430
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCL2 PubMed Links

Gene Details

Gene
BCL2
Gene Name
BCL2, apoptosis regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000633.2 4277 UTR 3 NP_000624.2
NM_000657.2 4277 Intron NP_000648.2
XM_011526135.2 4277 Intron XP_011524437.1
XM_017025917.1 4277 Intron XP_016881406.1

View Full Product Details