Product Details

SNP ID
rs7994795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:95804462 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTATAGAAACAGGAAAACTCAT[C/T]CTGAAATTCATACAGAATCACAAGG
Phenotype
MIM: 601184 MIM: 605898
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC3 PubMed Links

Gene Details

Gene
DNAJC3
Gene Name
DnaJ heat shock protein family (Hsp40) member C3
There are no transcripts associated with this gene.

Gene
UGGT2
Gene Name
UDP-glucose glycoprotein glucosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020121.3 Intron NP_064506.3
XM_011521094.1 Intron XP_011519396.1
XM_011521095.1 Intron XP_011519397.1
XM_011521096.1 Intron XP_011519398.1
XM_011521097.1 Intron XP_011519399.1
XM_011521098.1 Intron XP_011519400.1
XM_011521099.2 Intron XP_011519401.1
XM_011521100.2 Intron XP_011519402.1
XM_011521102.2 Intron XP_011519404.1
XM_011521103.2 Intron XP_011519405.1
XM_017020662.1 Intron XP_016876151.1
XM_017020663.1 Intron XP_016876152.1

View Full Product Details