Product Details

SNP ID
rs8191525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11770882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTAGCTATGCCGGGCTTACCTAC[G/T]TGGCTGGGGGTCTCACTTATGATTC
Phenotype
MIM: 608933
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C8orf49 PubMed Links

Gene Details

Gene
C8orf49
Gene Name
chromosome 8 open reading frame 49
There are no transcripts associated with this gene.

Gene
NEIL2
Gene Name
nei like DNA glycosylase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135746.1 Intron NP_001129218.1
NM_001135747.1 Intron NP_001129219.1
NM_001135748.1 Intron NP_001129220.1
NM_145043.2 Intron NP_659480.1
XM_005272382.2 Intron XP_005272439.1
XM_005272383.2 Intron XP_005272440.1
XM_017013301.1 Intron XP_016868790.1

View Full Product Details