Product Details

SNP ID
rs9683118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:142818839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAACACCAGGAAGCTGGACTGAGT[A/G]GTTAGATCAGCCATGTCCCGCTCTG
Phenotype
MIM: 607064
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCOLCE2 PubMed Links

Gene Details

Gene
PCOLCE2
Gene Name
procollagen C-endopeptidase enhancer 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013363.3 Intron NP_037495.1

View Full Product Details