Product Details

SNP ID
rs9299202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97244255 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAAAAAGCAACAGAATGGTAAGC[C/T]GAAAGGTCTGGTTATTGTCAGGGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD18CP PubMed Links

Gene Details

Gene
ANKRD18CP
Gene Name
ankyrin repeat domain 18C, pseudogene
There are no transcripts associated with this gene.

Gene
LOC100499484
Gene Name
SUGT1-1300002K09Rik pseudogene
There are no transcripts associated with this gene.

Gene
LOC100499484-C9ORF174
Gene Name
LOC100499484-C9orf174 readthrough
There are no transcripts associated with this gene.

View Full Product Details