Product Details
- SNP ID
-
rs9938930
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:50175336 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTTTCTTTGATGGATTTTGCTTT[G/T]GGTATTGTGTCTAGAAAGTCTTCGC
- Phenotype
-
MIM: 605540
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PAPD5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75238569] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PAPD5
- Gene Name
- PAP associated domain containing 5
View Full Product Details