Product Details

SNP ID
rs10162006
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113980420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCACCACCTCCTCCCACGTGTTC[A/G]CCTCCTCCGTGTGTGCACCTCCTGC
Phenotype
MIM: 605182
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LOC100507747 PubMed Links
Additional Information
For this assay, SNP(s) [rs117234914] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LOC100507747
Gene Name
uncharacterized LOC100507747
There are no transcripts associated with this gene.

Gene
RASA3
Gene Name
RAS p21 protein activator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320821.1 Intron NP_001307750.1
NM_001320822.1 Intron NP_001307751.1
NM_007368.3 Intron NP_031394.2
XM_011534841.2 Intron XP_011533143.1
XM_011534842.2 Intron XP_011533144.1
XM_017020438.1 Intron XP_016875927.1
XM_017020439.1 Intron XP_016875928.1

View Full Product Details