Product Details
- SNP ID
-
rs6060453
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31258983 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACACACACAAACACAACTTCAGC[C/T]TGCAGCTCAGAAATGTAAATTGGTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB115
PubMed Links
Gene Details
- Gene
- DEFB115
- Gene Name
- defensin beta 115
View Full Product Details