Product Details

SNP ID
rs6119719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31753272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACATTGTGCTGAAGTCACGAGC[C/T]TCAGGTCCAGATCTGGCTCTGTTTT
Phenotype
MIM: 605917
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TPX2 PubMed Links

Gene Details

Gene
TPX2
Gene Name
TPX2, microtubule nucleation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012112.4 Intron NP_036244.2
XM_011528697.2 Intron XP_011526999.1
XM_011528699.2 Intron XP_011527001.1
XM_011528700.2 Intron XP_011527002.1

View Full Product Details