Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001013627.2 | Intron | NP_001013649.2 | |||
XM_011530932.1 | Intron | XP_011529234.1 | |||
XM_011530933.1 | Intron | XP_011529235.1 | |||
XM_011530934.2 | Intron | XP_011529236.1 | |||
XM_017029473.1 | Intron | XP_016884962.1 | |||
XM_017029474.1 | Intron | XP_016884963.1 | |||
XM_017029475.1 | Intron | XP_016884964.1 | |||
XM_017029476.1 | Intron | XP_016884965.1 | |||
XM_017029477.1 | Intron | XP_016884966.1 |