Product Details

SNP ID
rs9457535
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:158948255 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGGCTGAGGGAGTTCAATGGGACT[A/G]TGTTAACTGACATATTCCAATAGAA
Phenotype
MIM: 615876
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RSPH3 PubMed Links

Gene Details

Gene
RSPH3
Gene Name
radial spoke 3 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031924.4 Intron NP_114130.3
XM_005267153.4 Intron XP_005267210.1
XM_017011347.1 Intron XP_016866836.1

View Full Product Details