Product Details

SNP ID
rs9825184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:15167646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCGTCAGAGAAAAGGACCACC[A/G]TGCAGGAAAGGGCATCTCTGCAGCT
Phenotype
MIM: 612397
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL6A4P1 PubMed Links

Gene Details

Gene
COL6A4P1
Gene Name
collagen type VI alpha 4 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details