Product Details

SNP ID
rs4982403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21067372 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAAACAAATTTAACTTTTTTAGC[C/T]TGATGGTTTGGTGGCAGAAATGGTG
Phenotype
MIM: 610018 MIM: 605272 MIM: 612485
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF40 PubMed Links

Gene Details

Gene
ARHGEF40
Gene Name
Rho guanine nucleotide exchange factor 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278529.1 Intron NP_001265458.1
NM_001278530.1 Intron NP_001265459.1
NM_018071.4 Intron NP_060541.3
XM_005267844.2 Intron XP_005267901.1
XM_011536937.2 Intron XP_011535239.1
XM_011536938.2 Intron XP_011535240.1
XM_017021434.1 Intron XP_016876923.1
XM_017021435.1 Intron XP_016876924.1
XM_017021436.1 Intron XP_016876925.1
XM_017021437.1 Intron XP_016876926.1
Gene
NDRG2
Gene Name
NDRG family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282211.1 Intron NP_001269140.1
NM_001282212.1 Intron NP_001269141.1
NM_001282213.1 Intron NP_001269142.1
NM_001282214.1 Intron NP_001269143.1
NM_001282215.1 Intron NP_001269144.1
NM_001282216.1 Intron NP_001269145.1
NM_001320329.1 Intron NP_001307258.1
NM_016250.2 Intron NP_057334.1
NM_201535.1 Intron NP_963293.1
NM_201536.1 Intron NP_963294.1
NM_201537.1 Intron NP_963831.1
NM_201538.1 Intron NP_963832.1
NM_201539.1 Intron NP_963833.1
NM_201540.1 Intron NP_963834.1
NM_201541.1 Intron NP_963835.1
XM_011536996.2 Intron XP_011535298.1
XM_011536997.1 Intron XP_011535299.1
XM_011536998.1 Intron XP_011535300.1
XM_011536999.1 Intron XP_011535301.1
XM_011537001.1 Intron XP_011535303.1
XM_011537002.1 Intron XP_011535304.1
XM_017021480.1 Intron XP_016876969.1
XM_017021481.1 Intron XP_016876970.1
XM_017021482.1 Intron XP_016876971.1
XM_017021483.1 Intron XP_016876972.1
XM_017021484.1 Intron XP_016876973.1
XM_017021485.1 Intron XP_016876974.1
XM_017021486.1 Intron XP_016876975.1
XM_017021487.1 Intron XP_016876976.1
XM_017021488.1 Intron XP_016876977.1
XM_017021489.1 Intron XP_016876978.1
XM_017021490.1 Intron XP_016876979.1
XM_017021491.1 Intron XP_016876980.1
XM_017021492.1 Intron XP_016876981.1
XM_017021493.1 Intron XP_016876982.1
XM_017021494.1 Intron XP_016876983.1
XM_017021495.1 Intron XP_016876984.1
XM_017021496.1 Intron XP_016876985.1
XM_017021497.1 Intron XP_016876986.1
Gene
RNASE8
Gene Name
ribonuclease A family member 8
There are no transcripts associated with this gene.

View Full Product Details