Product Details

SNP ID
rs10149038
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:55130387 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCTGGGAATTGGGTTGCTGCGCA[A/G]TGAGGACACCTGGACCTCAGTGACT
Phenotype
MIM: 153619
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LGALS3 PubMed Links

Gene Details

Gene
LGALS3
Gene Name
lectin, galactoside binding soluble 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177388.1 Intron NP_001170859.1
NM_002306.3 Intron NP_002297.2
XM_011536759.2 Intron XP_011535061.2

View Full Product Details