Product Details

SNP ID
rs7683615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:184697219 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTAAAATCCTACCTACCACTTATA[C/T]GGGAAAATAGCTTGTTTTTCATATG
Phenotype
MIM: 611511 MIM: 615421
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPU PubMed Links

Gene Details

Gene
CENPU
Gene Name
centromere protein U
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024629.3 Intron NP_078905.2
XM_005263218.4 Intron XP_005263275.2
Gene
PRIMPOL
Gene Name
primase and DNA directed polymerase
There are no transcripts associated with this gene.

View Full Product Details