Product Details

SNP ID
rs6566813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:74540552 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATGTGACATAAGAGGGAATGGCA[A/G]CAGCTCTCTTGGGGAACCTCAGTGA
Phenotype
MIM: 609064
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNDP1 PubMed Links

Gene Details

Gene
CNDP1
Gene Name
carnosine dipeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032649.5 Intron NP_116038.4

View Full Product Details