Product Details

SNP ID
rs9879386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172507439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGAGTCAAAGGCACATCCAAGC[A/G]GATGGCGTCTCTAGCCAAAAATAAT
Phenotype
MIM: 603598
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFSF10 PubMed Links

Gene Details

Gene
TNFSF10
Gene Name
tumor necrosis factor superfamily member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190942.1 Intron NP_001177871.1
NM_001190943.1 Intron NP_001177872.1
NM_003810.3 Intron NP_003801.1

View Full Product Details