Product Details

SNP ID
rs9481825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118554929 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAGTACAACTTACCAGTGTCAAC[A/G]TAGGGGATGACTCCTGTATATGCTT
Phenotype
MIM: 172405
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP85L PubMed Links

Gene Details

Gene
CEP85L
Gene Name
centrosomal protein 85 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042475.2 Intron NP_001035940.1
NM_001178035.1 Intron NP_001171506.1
NM_206921.2 Intron NP_996804.2
XM_005266970.1 Intron XP_005267027.1
XM_005266971.1 Intron XP_005267028.1
XM_005266972.4 Intron XP_005267029.1
XM_006715475.3 Intron XP_006715538.1
XM_011535809.2 Intron XP_011534111.1
XM_011535810.2 Intron XP_011534112.1
XM_011535811.1 Intron XP_011534113.1
XM_017010846.1 Intron XP_016866335.1
XM_017010847.1 Intron XP_016866336.1
Gene
PLN
Gene Name
phospholamban
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002667.4 Intron NP_002658.1

View Full Product Details