Product Details

SNP ID
rs5960058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:54942547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCCAGGACTGCTTAGAAGAAG[C/G]AAATGAAATCTCTCTGGATTCTCTC
Phenotype
MIM: 311790
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PFKFB1 PubMed Links

Gene Details

Gene
PFKFB1
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271804.1 Intron NP_001258733.1
NM_001271805.1 Intron NP_001258734.1
NM_002625.3 Intron NP_002616.2
XM_017029576.1 Intron XP_016885065.1
XM_017029577.1 Intron XP_016885066.1
XM_017029578.1 Intron XP_016885067.1

View Full Product Details