Product Details

SNP ID
rs10409854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1756940 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCGATACCCACCACCTCCTGCCTC[C/T]TCTCTAGCACATATTTGGGGGACAT
Phenotype
MIM: 611294
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ONECUT3 PubMed Links

Gene Details

Gene
ONECUT3
Gene Name
one cut homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080488.1 Intron NP_001073957.1

View Full Product Details