Product Details

SNP ID
rs9381196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42570065 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGTTAGCATATGTTAGAACTAA[C/T]ATGAAAAGCTATAGATTTTATTCTT
Phenotype
MIM: 609134
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBR2 PubMed Links

Gene Details

Gene
UBR2
Gene Name
ubiquitin protein ligase E3 component n-recognin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184801.1 Intron NP_001171730.1
NM_015255.2 Intron NP_056070.1
XM_005248965.4 Intron XP_005249022.1
XM_005248966.3 Intron XP_005249023.1
XM_011514438.2 Intron XP_011512740.2
XM_011514439.1 Intron XP_011512741.1
XM_011514440.1 Intron XP_011512742.1
XM_011514441.2 Intron XP_011512743.1
XM_017010594.1 Intron XP_016866083.1
XM_017010595.1 Intron XP_016866084.1
XM_017010596.1 Intron XP_016866085.1
XM_017010597.1 Intron XP_016866086.1

View Full Product Details