Product Details
- SNP ID
-
rs6117978
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:7886586 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAATGCAAACACAACTAATAATAA[A/G]ACAACTTAGTCTCTTATTTTCTCAT
- Phenotype
-
MIM: 605023
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HAO1
PubMed Links
Gene Details
- Gene
- HAO1
- Gene Name
- hydroxyacid oxidase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017545.2 |
|
Intron |
|
|
NP_060015.1 |
View Full Product Details