Product Details

SNP ID
rs10203236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:63051552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAGTGCTAAGTTTGGAGAAAGT[A/C]TTTGAAAACTCACGCTGCAGCTTCG
Phenotype
MIM: 609922 MIM: 600036
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EHBP1 PubMed Links

Gene Details

Gene
EHBP1
Gene Name
EH domain binding protein 1
There are no transcripts associated with this gene.

Gene
LOC100132215
Gene Name
uncharacterized LOC100132215
There are no transcripts associated with this gene.

Gene
OTX1
Gene Name
orthodenticle homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199770.1 Intron NP_001186699.1
NM_014562.3 Intron NP_055377.1

View Full Product Details