Product Details
- SNP ID
-
hCV30017088
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:40349333 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGAGGTCTCAGGCAGAAGGAACAG[A/T]CAAGAGCAGGATTATGTTGTTGTTT
- Phenotype
-
MIM: 608222
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADSL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77810402] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADSL
- Gene Name
- adenylosuccinate lyase
There are no transcripts associated with this gene.
View Full Product Details