Product Details
- SNP ID
-
hCV30018082
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31752954 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTGTGAAGATATAGAGTGTCATC[C/T]TTGAATGTTACTGAGAAACATTCAA
- Phenotype
-
MIM: 605917
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TPX2
PubMed Links
Gene Details
- Gene
- TPX2
- Gene Name
- TPX2, microtubule nucleation factor
There are no transcripts associated with this gene.
View Full Product Details