Product Details
- SNP ID
-
hCV30019053
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:17409372 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTAGTTTCCATGTGACTGCCACCAA[C/T]GCTCTGCCAATCAGAGCCTTCCAGG
- Phenotype
-
MIM: 610363
MIM: 609587
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PADI6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115730509] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PADI6
- Gene Name
- peptidyl arginine deiminase 6
There are no transcripts associated with this gene.
- Gene
- RCC2
- Gene Name
- regulator of chromosome condensation 2
There are no transcripts associated with this gene.
View Full Product Details