Product Details
- SNP ID
-
rs9652843
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41636711 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCGTGGGGCCACTCCCTGGCCAGGG[C/G]TAGGAAAATGCAAACATTCCTCCTG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT42P
PubMed Links
Gene Details
- Gene
- KRT42P
- Gene Name
- keratin 42 pseudogene
There are no transcripts associated with this gene.
View Full Product Details