Product Details

SNP ID
rs9928139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1942811 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGTAGGGAAGGGGTACCACCCCC[A/T]CGTATCCCGATCCCTTAGACCCACC
Phenotype
MIM: 606216
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MSRB1 PubMed Links

Gene Details

Gene
MSRB1
Gene Name
methionine sulfoxide reductase B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016332.2 Intron NP_057416.1
Gene
RPL3L
Gene Name
ribosomal protein L3 like
There are no transcripts associated with this gene.

View Full Product Details