Product Details

SNP ID
rs10197007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:203947637 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTATTGTTAGCAGCAGTTAACAG[G/T]CTTGAGCCTCTACTGGGAAGAATAT
Phenotype
MIM: 604558
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ICOS PubMed Links

Gene Details

Gene
ICOS
Gene Name
inducible T-cell costimulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012092.3 Intron NP_036224.1

View Full Product Details