Product Details

SNP ID
hCV30043522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:59810 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAATTCTAGAAATTCAGTGATGT[A/C]AAATATTGTTTTCCACATCTAAAAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF595 PubMed Links

Gene Details

Gene
ZNF595
Gene Name
zinc finger protein 595
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286052.1 Intron NP_001272981.1
NM_001286053.1 Intron NP_001272982.1
NM_001286054.1 Intron NP_001272983.1
NM_182524.3 Intron NP_872330.1

View Full Product Details